ENA390552219 Enamine

2-(aminomethyl)-5,6,7,8-tetrahydroquinolin-5-one dihydrochloride

  • MDL number MFCD24369200
  • Cas Number : 1432681-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127416

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127416
Availability details for
Quantity
  • Availability
  • Add to Cart