ENA390552220 Enamine

N-(1-benzyl-4-cyanopiperidin-4-yl)-2,2,2-trifluoroacetamide; trifluoroacetic acid

  • MDL number MFCD24369201
  • Cas Number : 1432682-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208...210
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127418

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127418
Availability details for
Quantity
  • Availability
  • Add to Cart