ENA390552221 Enamine

5-(aminomethyl)-3-methyl-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD24369202
  • Cas Number : 1432679-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127488

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127488
Availability details for
Quantity
  • Availability
  • Add to Cart