ENA390552226 Enamine

2-methoxyethyl 2-(2-methoxyethoxy)pyridine-4-carboxylate

  • MDL number MFCD24369208
  • Cas Number : 1432680-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127556

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127556
Availability details for
Quantity
  • Availability
  • Add to Cart