ENA390552228 Enamine

3-hydroxy-2,2-dimethylcyclobutan-1-one

  • MDL number MFCD24369210
  • Cas Number : 1432681-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127594

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127594
Availability details for
Quantity
  • Availability
  • Add to Cart