ENA390552235 Enamine

6-chloro-N-methyl-2,3,4,9-tetrahydro-1H-carbazol-3-amine hydrochloride

  • MDL number MFCD24369218
  • Cas Number : 1432679-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127653
Availability details for
Quantity
  • Availability
  • Add to Cart