ENA390552237 Enamine

5-(benzyloxy)-3,4-dihydroquinazolin-4-one

  • MDL number MFCD24369220
  • Cas Number : 1432681-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127661
Availability details for
Quantity
  • Availability
  • Add to Cart