ENA390552238 Enamine

1-[(2-nitrophenyl)methyl]imidazolidin-2-one

  • MDL number MFCD24189886
  • Cas Number : 1432678-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127667
Availability details for
Quantity
  • Availability
  • Add to Cart