ENA390552239 Enamine

N-(9-fluoro-2,3,4,5-tetrahydro-1-benzoxepin-5-ylidene)hydroxylamine

  • Cas Number : 1432684-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127678
Availability details for
Quantity
  • Availability
  • Add to Cart