ENA390552241 Enamine

2-cyclobutylideneethan-1-ol

  • MDL number MFCD24369225
  • Cas Number : 4415-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127684
Availability details for
Quantity
  • Availability
  • Add to Cart