ENA390552246 Enamine

N-methyl-N-(2,2,2-trifluoroethyl)piperidin-4-amine hydrochloride

  • MDL number MFCD24369230
  • Cas Number : 1432678-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127734
Availability details for
Quantity
  • Availability
  • Add to Cart