ENA390552253 Enamine

[3-(2-methoxynaphthalen-1-yl)-1,2-oxazol-5-yl]methanamine hydrochloride

  • MDL number MFCD24369237
  • Cas Number : 1432681-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127844

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127844
Availability details for
Quantity
  • Availability
  • Add to Cart