ENA390552254 Enamine

tert-butyl N-[3-methoxy-1-(5-methylfuran-2-yl)propyl]carbamate

  • MDL number MFCD24369239
  • Cas Number : 1432680-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127865
Availability details for
Quantity
  • Availability
  • Add to Cart