ENA390552260 Enamine

2-amino-5-(4-methylphenyl)-4,5-dihydro-1,3-oxazol-4-one dihydrochloride

  • MDL number MFCD24369245
  • Cas Number : 1432680-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127915
Availability details for
Quantity
  • Availability
  • Add to Cart