ENA390552263 Enamine

2-{[(4-methoxyphenyl)methyl]amino}-3-methylbutan-1-ol hydrochloride

  • MDL number MFCD24369249
  • Cas Number : 1432682-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127927
Availability details for
Quantity
  • Availability
  • Add to Cart