ENA390552265 Enamine

[4-methyl-5-(propan-2-yl)-4H-1,2,4-triazol-3-yl]methanamine dihydrochloride

  • MDL number MFCD24369253
  • Cas Number : 1432680-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127966

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127966
Availability details for
Quantity
  • Availability
  • Add to Cart