ENA390552268 Enamine

1-(5-hydroxy-2,3,4,5-tetrahydro-1H-2-benzazepin-2-yl)ethan-1-one

  • MDL number MFCD24369256
  • Cas Number : 1432680-96-0
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(5-hydroxy-1,3,4,5-tetrahydro-2H-benzo[c]azepin-2-yl)ethan-1-one
Inchi Code 1S/C12H15NO2/c1-9(14)13-7-6-12(15)11-5-3-2-4-10(11)8-13/h2-5,12,15H,6-8H2,1H3
Inchi Key FHFPBWBQLUPNJF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128007
Availability details for
Quantity
  • Availability
  • Add to Cart