ENA390552345 Enamine

3-methylquinolin-4-amine

  • MDL number MFCD01646081
  • Cas Number : 19701-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126795
Availability details for
Quantity
  • Availability
  • Add to Cart