ENA390552346 Enamine

1-(propan-2-yl)-2,3,4,5-tetrahydro-1H-1,4-benzodiazepine

  • MDL number MFCD18268858
  • Cas Number : 1267815-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-126796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126796
Availability details for
Quantity
  • Availability
  • Add to Cart