ENA390552348 Enamine

5-[(3-fluorophenyl)methyl]-1H-1,2,4-triazol-3-amine

  • MDL number MFCD14610757
  • Cas Number : 627544-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126799

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-126799
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart