ENA390552354 Enamine

(5-cyclopropyl-4-methyl-4H-1,2,4-triazol-3-yl)methanamine dihydrochloride

  • MDL number MFCD24369132
  • Cas Number : 1432681-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126805

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126805
Availability details for
Quantity
  • Availability
  • Add to Cart