ENA390552355 Enamine

[1-(5-bromo-7-methyl-1-benzofuran-2-yl)-2-methylpropyl](methyl)amine hydrochloride

  • MDL number MFCD24369133
  • Cas Number : 1432681-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126808
Availability details for
Quantity
  • Availability
  • Add to Cart