ENA390552380 Enamine

3-fluoro-2-nitrobenzene-1-sulfonamide

  • MDL number MFCD16044947
  • Cas Number : 1251363-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-126884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126884
Availability details for
Quantity
  • Availability
  • Add to Cart