ENA390552384 Enamine

7-hydroxynaphthalen-2-yl acetate

  • MDL number MFCD24369086
  • Cas Number : 146744-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122601

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122601
Availability details for
Quantity
  • Availability
  • Add to Cart