ENA390552385 Enamine

3-(3,5-dimethyl-1H-pyrazol-4-yl)morpholine

  • MDL number MFCD22064708
  • Cas Number : 1367699-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122665
Availability details for
Quantity
  • Availability
  • Add to Cart