ENA390552386 Enamine

6-methanesulfonyl-2-(pyridin-2-yl)-1H-1,3-benzodiazole

  • MDL number MFCD17296486
  • Cas Number : 1256094-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126892

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126892
Availability details for
Quantity
  • Availability
  • Add to Cart