ENA390552387 Enamine

2-(2-cyanophenoxy)benzoic acid

  • MDL number MFCD11529197
  • Cas Number : 209461-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126895

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-126895
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart