ENA390552389 Enamine

2-ethylquinolin-4-amine

  • MDL number MFCD16690993

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-122815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122815
Availability details for
Quantity
  • Availability
  • Add to Cart