ENA390552391 Enamine

5,8-dichloroquinolin-2-ol

  • MDL number MFCD19678294
  • Cas Number : 1343820-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126921

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126921
Availability details for
Quantity
  • Availability
  • Add to Cart