ENA390552395 Enamine

2-(4-amino-2,3-dihydro-1H-isoindol-2-yl)ethan-1-ol

  • MDL number MFCD10007254
  • Cas Number : 1017388-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126925
Availability details for
Quantity
  • Availability
  • Add to Cart