ENA390552402 Enamine

4-(trichloroacetyl)-1H-pyrrole-2-carbonitrile

  • MDL number MFCD19981427
  • Cas Number : 66832-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-126936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126936
Availability details for
Quantity
  • Availability
  • Add to Cart