ENA390552408 Enamine

oxepan-4-amine

  • MDL number MFCD16038648
  • Cas Number : 911825-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126949
Availability details for
Quantity
  • Availability
  • Add to Cart