ENA390552410 Enamine

5-cyclopropyl-1-methyl-1H-pyrazol-4-amine

  • MDL number MFCD21773853
  • Cas Number : 1393100-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126952
Availability details for
Quantity
  • Availability
  • Add to Cart