ENA390552416 Enamine

[2-(benzylsulfanyl)phenyl]methanol

  • MDL number MFCD01830350
  • Cas Number : 4521-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126963
Availability details for
Quantity
  • Availability
  • Add to Cart