ENA390552423 Enamine

1-[4-(ethanesulfonyl)phenyl]methanamine

  • MDL number MFCD10032480
  • Cas Number : 583837-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126973
Availability details for
Quantity
  • Availability
  • Add to Cart