ENA390552427 Enamine

2-(oxan-4-yl)pyrrolidine hydrochloride

  • MDL number MFCD24369153
  • Cas Number : 1432679-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126977
Availability details for
Quantity
  • Availability
  • Add to Cart