ENA390552429 Enamine

[2-(butan-2-yloxy)pyridin-4-yl]methanamine

  • MDL number MFCD16043918
  • Cas Number : 1247570-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126981

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126981
Availability details for
Quantity
  • Availability
  • Add to Cart