ENA390552432 Enamine

1-(3,4-dichlorophenyl)-5-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08443789
  • Cas Number : 1020724-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127021

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127021
Availability details for
Quantity
  • Availability
  • Add to Cart