ENA390552436 Enamine

methyl[(3,4,5-trifluorophenyl)methyl]amine

  • MDL number MFCD17284527
  • Cas Number : 1340036-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127025

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127025
Availability details for
Quantity
  • Availability
  • Add to Cart