ENA390552447 Enamine

3-amino-1-{[3-(trifluoromethyl)phenyl]methyl}piperidin-2-one

  • MDL number MFCD19099561
  • Cas Number : 1404945-84-1
  • Molecular Weight : 272.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127049
Availability details for
Quantity
  • Availability
  • Add to Cart