ENA390552449 Enamine

2-bromo-3,6-dimethylpyridine

  • MDL number MFCD12400995
  • Cas Number : 38749-92-7
  • Molecular Weight : 186.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-3,6-dimethylpyridine
Inchi Code 1S/C7H8BrN/c1-5-3-4-6(2)9-7(5)8/h3-4H,1-2H3
Inchi Key BKLNQHXCFLTIJA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127051

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-127051
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart