ENA390552451 Enamine

[4-(butan-2-yl)phenyl]methanol

  • MDL number MFCD16301138
  • Cas Number : 102934-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-127053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127053
Availability details for
Quantity
  • Availability
  • Add to Cart