ENA390552455 Enamine

5-chloro-8-methylquinolin-2-ol

  • MDL number MFCD19678298
  • Cas Number : 73108-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254...256
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127062

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127062
Availability details for
Quantity
  • Availability
  • Add to Cart