ENA390552456 Enamine

2-(butan-2-yloxy)pyridine-4-carboxylic acid

  • MDL number MFCD14588116
  • Cas Number : 1250383-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83...85
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127063

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127063
Availability details for
Quantity
  • Availability
  • Add to Cart