ENA390552460 Enamine

3-ethyl-6-methyl-4-oxo-3H,4H-furo[2,3-d]pyrimidine-5-carboxylic acid

  • MDL number MFCD09880378
  • Cas Number : 1023815-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127067

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127067
Availability details for
Quantity
  • Availability
  • Add to Cart