ENA390552466 Enamine

1-(3,5-dimethylphenyl)-5-methyl-1H-pyrazol-3-amine

  • MDL number MFCD16845351
  • Cas Number : 1249554-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127075
Availability details for
Quantity
  • Availability
  • Add to Cart