ENA390552467 Enamine

2-(2-methoxyethoxy)pyridin-4-amine

  • MDL number MFCD13190649
  • Cas Number : 936112-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127076

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127076
Availability details for
Quantity
  • Availability
  • Add to Cart