ENA390552468 Enamine

7-bromo-8-(chloromethyl)-3,4-dihydro-2H-1,5-benzodioxepine

  • MDL number MFCD06496438
  • Cas Number : 893722-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127077

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127077
Availability details for
Quantity
  • Availability
  • Add to Cart