ENA390552470 Enamine

methyl 1-methyl-1H-indole-7-carboxylate

  • MDL number MFCD12025398
  • Cas Number : 167479-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127079
Availability details for
Quantity
  • Availability
  • Add to Cart