ENA390552479 Enamine

1-(3,4-dichlorophenyl)-5-methyl-1H-pyrazol-3-amine

  • MDL number MFCD14237201
  • Cas Number : 1172560-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127094
Availability details for
Quantity
  • Availability
  • Add to Cart